Honeywell Atlas Fit Black Natural Rubber-Coated Gloves

For greater comfort and productivity

Manufacturer: Honeywell 320PC-M

 View all options for this product

Catalog No. 19-047-705


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Coated Area Palm, finger
    Color Gray, black
    Cut Resistance Rating Level 1
    Cleaning Procedure Withstands commercial laundry room detergents (up to pH 14) and warm water temperatures up to 120°F. Lower heat settings up to 180°F are suggested for drying as extreme heat may cause shrinking and degradation.
    Exterior Finish Rough
    Glove Cuff Style Knit
    Glove Cuff Color Blue
    For Use With (Application) construction, recycling, lumber and mill work, Glass handling, Metal stamping, automotive, landscaping
    Material Natural Rubber
    Liner Material Cotton, Polyester
    Size Medium

    Seamless knit cotton/polyester blend shell has a natural rubber coating on the palm that resists abrasion, protecting the hand from sharp objects.

    • Textured palm coating provides a safe, sure grip while inhibiting liquid penetration
    • Fingertips and thumb crotch are also rubber coated to minimize wear in these critical areas, increasing glove life
    • Ergonomically shaped to conform snugly to the hand, reducing hand fatigue
    • Dark color hides dirt and grime

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit