Learn More
Honeywell™ B-Safe Lockout/Tagout Devices for Valves
Manufacturer:  Honeywell™ BS01G
 View more versions of this product
Catalog No. 19-170-182
Add to cart
Specifications
Specifications
Lockout/Tagout Device
To prevent accidental activation
-50° to 360°F
0.87 in.
Polypropylene
Green
2 to 8 in. ball valve
-46° to 182°C
2cm