Learn More
Honeywell™ B-Safe Lockout/Tagout Devices for Valves
Supplier: Honeywell™ BS02B
Description
Specifications
Lockout/Tagout Device | |
Polypropylene | |
0.78 cm | |
1.5 to 2.5 in ball valve | |
−46° to +182°C |
Blue | |
0.31 in. | |
Locking out quarter turn ball valves with flat 'dog-leg' handles | |
−50° to +360°F |