Learn More
Honeywell™ B-Safe Lockout/Tagout Devices for Valves
Manufacturer:  Honeywell™ BS04B
 View more versions of this product
Catalog No. 19-170-193
Add to cart
Specifications
Specifications
Lockout/Tagout Device
0.37 in.
Manufacturing, mining, municipal services, oil and gas, utilities
-50° to 360°F
1.62 in.
Polypropylene
Blue
0.93cm
Butterfly valve
-46° to 182°C
4cm