Learn More
Honeywell™ B-Safe Lockout/Tagout Devices for Valves
Supplier: Honeywell™ BS03B
Description
Specifications
Lockout/Tagout Device | |
Polypropylene | |
0.78 cm | |
2 to 8 in. ball valve | |
−46° to +182°C | |
4 cm |
Blue | |
0.31 in. | |
Manufacturing, mining, municipal services, oil and gas, utilities | |
−50° to +360°F | |
1.62 in. |