Learn More
Honeywell B-Safe Lockout/Tagout Devices for Valves
Manufacturer:  Honeywell BS03Y
 View more versions of this product
Catalog No. 19-170-191
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Polypropylene
Yellow
0.78cm
2 to 8 in. ball valve
-46° to 182°C
4cm
Lockout/Tagout Device
0.31 in.
Manufacturing, mining, municipal services, oil and gas, utilities
-50° to 360°F
1.62 in.