Learn More
Honeywell B-Safe Lockout/Tagout Devices for Valves
Manufacturer:  Honeywell BS01B
 View more versions of this product
Catalog No. 19-170-181
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Blue
2 to 8 in. ball valve
-46° to 182°C
2cm
Lockout/Tagout Device
To prevent accidental activation
-50° to 360°F
0.87 in.
Polypropylene