Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Barricade Tape
Dispenses quickly and easily to effectively designate a hazardous area
Supplier: Honeywell™ CT3YE2
Description
- Polyethylene tape with repeating message (Danger Do Not Enter, Caution Do Not Enter, Caution Buried Cable, Danger or Caution)
- Width: 7.62cm (3 in.)
- Comes in convenient dispenser box
Specifications
Yellow | |
3 in. | |
HDPE | |
Caution Do Not Enter | |
1000 ft. | |
Barricade Tape |
7.62 cm | |
0.002 mil | |
1000 ft. x 3 in. (304.8 m x 7.62 cm) | |
Dispenses quickly and easily to effectively designate a hazardous area | |
304.8 m |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title