Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Barricade Tape
Dispenses quickly and easily to effectively designate a hazardous area
$27.45
Specifications
Material | HDPE |
---|---|
Product Type | Barricade Tape |
For Use With (Application) | Dispenses quickly and easily to effectively designate a hazardous area |
Thickness | 0.002 mil |
Width (English) | 3 in. |
Catalog Number | Mfr. No. | Color | Displayed Text | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Displayed Text | Length (Metric) | Price | Quantity & Availability | |||
19-170-115
|
Honeywell
CT3YE2 |
Yellow | Caution Do Not Enter | 304.8 m |
Each for $27.45
|
|
19-120-630
|
Honeywell
CT3YE1/300N |
Yellow | Caution | 91.4 m |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-120-633
|
Honeywell
CT3RE9/300N |
Red | Caution | 91.4 m |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-120-629
|
Honeywell
CT3RE9 |
Red | Danger | 304.8 m |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-120-632
|
Honeywell
CT3RE8/300N |
Red | Danger Do Not Enter | 91.4 m |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-170-117
|
Honeywell
CT3YE1 |
Yellow | Caution | 304.8 m |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Polyethylene tape with repeating message (Danger Do Not Enter, Caution Do Not Enter, Caution Buried Cable, Danger or Caution)
- Width: 7.62cm (3 in.)
- Comes in convenient dispenser box
Specifications
HDPE | |
Dispenses quickly and easily to effectively designate a hazardous area | |
3 in. |
Barricade Tape | |
0.002 mil | |
7.62 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title