Honeywell Barricade Tape

Dispenses quickly and easily to effectively designate a hazardous area

Manufacturer: Honeywell CT3RE8

 View more versions of this product

Catalog No. 19-170-116

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Dispenses quickly and easily to effectively designate a hazardous area
    Thickness 0.002mil
    Product Type Barricade Tape
    Length (English) 1000 ft.
    Length (Metric) 304.8m
    Width (English) 3 in.
    Width (Metric) 7.62cm
    Displayed Text Danger Do Not Enter
    Dimensions (L x W) 1000 ft. x 3 in. (304.8m x 7.62cm)
    Color Red
    Material Polyethylene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit