Honeywell Ben 2 Plus Structural Helmets Available on GSA/VA Contract for Federal Government customers only.
Rugged and dependable helmet for structural firefighting operations
Manufacturer: Total Fire Group SAEGSA00522
 View more versions of this product
Catalog No. 19-167-093
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance NFPA 1971, 2007 Edition.
    Liner Material Kevlar Felt Winter
    Color Yellow
    Material Shell: FYR-Glass; Ear Covers: Yellow Nomex FR Cotton
  • Description & Specifications
Related Products
Product Content Feedback