Honeywell Blue 1H Half Mask Respirators

Designed with healthcare workers for healthcare workers

Manufacturer: Honeywell B210070

 View more versions of this product

Catalog No. 19-066-241

  Check Availability
Add to Cart

Includes: One pair of P100 particulate filters and filter splash guards.

    Specifications

    For Use With (Application) Healthcare professionals in environments ranging from the ER, OR, and IC to clinics and other medical facilities
    Latex Free No
    Cartridge Type P100 particulate cartridge
    Color Light green
    Disposable No
    Facepiece Type Half mask
    Includes P100 filters and splash guards
    Material Silicone
    Suspension Type Cradle
    Product Type Half-face respirator
    Size Small

    Reusable respirator to protect healthcare workers from the hazards and challenges faced each day in infectious disease environments or during a pandemic

    • Includes a speech diaphragm for better worker-to-patient speech clarity for improved communication
    • Splash guards for the P100 filters help prevent blood borne pathogens from contaminating the exterior of the filters
    • Splash guard over the exhalation port directs healthcare worker's exhaled breath away from patients
    • Silicone facepiece is soft and flexible to provide workers with a comfortable fit for hours of use
    • Dual sealing flange provides excellent facepiece seal
    • Cradle suspension provides excellent support for a secure fit
    • NIOSH approved (with P100 particulate filters only)

    Order Info

    Available in light green and lilac colors of scrubs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit