Learn More
Honeywell Blue 1H Half Mask Respirators
Designed with healthcare workers for healthcare workers
Manufacturer:  Honeywell B230070
 View more versions of this product
Catalog No. 19-066-243
  Check Availability
Add to cart
Includes: One pair of P100 particulate filters and filter splash guards.
Specifications
Description
Reusable respirator to protect healthcare workers from the hazards and challenges faced each day in infectious disease environments or during a pandemic
  • Includes a speech diaphragm for better worker-to-patient speech clarity for improved communication
  • Splash guards for the P100 filters help prevent blood borne pathogens from contaminating the exterior of the filters
  • Splash guard over the exhalation port directs healthcare worker's exhaled breath away from patients
  • Silicone facepiece is soft and flexible to provide workers with a comfortable fit for hours of use
  • Dual sealing flange provides excellent facepiece seal
  • Cradle suspension provides excellent support for a secure fit
  • NIOSH approved (with P100 particulate filters only)
Order Info
Available in light green and lilac colors of scrubs.
Description & Specifications
Specifications
P100 particulate cartridge
No
Healthcare professionals in environments ranging from the ER, OR, and IC to clinics and other medical facilities
No
Cradle
Large
Light green
Half mask
P100 filters and splash guards
Silicone
Half-face respirator