Learn More
Honeywell BPR Bronze Structural Gear
Provide enhanced water repellence
Manufacturer:  Total Fire Group GSAPUS00040 XL
 View more versions of this product
Catalog No. 19-167-034D
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Bronze
X-Large
7.5 oz. PBI bronze Outer; Aralite/Nomex Thermal Liner; CrossTech/Nomex Moisture Barrier