Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Chemical Resistant Gloves

For cleanroom applications requiring good abrasion resistance as well as resistance to solvents

Manufacturer:  Honeywell SK142W/7/HEM

 View more versions of this product

Catalog No. 19-999-125


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Sand patch
Nitrile
15mil
Powder-free
13 in.
Straight
7
0120 CE, EN 420, EN 388, EN 374
White
33cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit