Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Dexi-Task Nitrile Gloves

For industrial examination and medical use

Manufacturer:  Honeywell LA049IND/S

 View more versions of this product

Catalog No. 19-150-1038

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Specifications

Textured
Yes
Ambidextrous
Yes
Small
3.3 in.
5 mil
Powdered
9 in.
9 x 3.3 in. (23 x 8.5cm)
Rolled
Industrial
Nitrile
16
8.5cm
USDA, FDA, 21 CFR, EN 455-1, 2, EN 374-1, EN 420, ASTM D 3578
Blue
23cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit