Honeywell Duro Task and Duro Task Plus Rubber Coated Gloves
Launderable work glove with natural rubber coating for a variety of applications
Manufacturer: Honeywell NF14/9L
 View more versions of this product
Catalog No. 19-150-0947
  Check Availability
Add to Cart
Alert
This product contains natural rubber latex proteins which may cause allergic reactions in some individuals.
    Specifications
    Cleaning Procedure Launderable
    Exterior Finish Textured
    Glove Cuff Style Knit
    Glove Cuff Color Brown
    For Use With (Application) automotive assembly, gardening, general maintenance work, janitorial, light assembly, manufacturing, small part handling, shipping/receiving
    Material Natural Rubber
    Liner Material Cotton, Polyester
    Size Large
    Coated Area Palm
    Color Blue, gray
    Cut Resistance Rating Level 4
  • Description & Specifications
Related Products
Product Content Feedback