Learn More
Honeywell DVO™ Laser Eyewear: Glendale XC
For use with low-powered lasers and by those exposed to diffuse laser radiation
Manufacturer:  Honeywell 3180103
 View more versions of this product
Catalog No. 19-150-0434
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Laser Eyewear
103
Orange
ANSI Z136.1 standards
Glendale XC Frame
24%