Honeywell DVO™ Laser Eyewear: Glendale XC
For use with low-powered lasers and by those exposed to diffuse laser radiation
Manufacturer: Honeywell 31-80101
 View more versions of this product
Catalog No. 19-150-0432
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance ANSI Z136.1 standards
    Filter Type 101
    Includes Glendale XC Frame
    Lens Tint Light Yellow-Green
    Transmittance Value 50%
    Type Laser Eyewear
  • Description & Specifications
Related Products
Product Content Feedback