Honeywell DVO™ Laser Eyewear: LOTG Series
For use with low-powered lasers and by those exposed to diffuse laser radiation
Manufacturer: Honeywell LOTGARGON/KTP
 View more versions of this product
Catalog No. 18-999-577B
  Check Availability
Add to Cart
    Specifications
    Type Laser Eyewear
    Certifications/Compliance ANSI Z136.1 standards
    Frame Material Nylon
    Includes LOTG Frame
    Lens Tint Orange
    Transmittance Value 53%
    Filters have excellent visible light transmission and will not degrade from UV exposure for a wide range of laser wavelengths (see specifications). Argon and helium-neon lenses let wearer see the termination point of the beam.
    • Universal design fits comfortably, even over prescription eyewear
    • Patented adjustable temples with Duoflex™ “comfort cushions” alleviate pressure
    • Nylon frame supports impact-resistant polycarbonate lens
    Do not use during exposure to direct laser radiation.
  • Description & Specifications
Related Products
Product Content Feedback