Honeywell DVO™ Laser Eyewear: LOTG Series
For use with low-powered lasers and by those exposed to diffuse laser radiation
Manufacturer: Honeywell LOTGPDT
 View more versions of this product
Catalog No. 18-999-580
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ANSI Z136.1 standards
    Frame Material Nylon
    Includes LOTG Frame
    Lens Tint Blue
    Transmittance Value 50%
    Type Laser Eyewear
  • Description & Specifications
Related Products
Product Content Feedback