Learn More
Honeywell E-Safe Electrical Lockout Devices
Manufacturer:  Honeywell LP550
 View more versions of this product
Catalog No. 19-170-202
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.375 in.
Manufacturing, mining, municipal services, oil and gas, utilities
2.99 in.
76mm
Sliding closure and integrated multiple lockout
25mm
76mm
Polypropylene
9.5mm
For 220/550V
3 in.
76mm
1 in.
3 in.
Red
Electrical Plug Lockout