Learn More
Honeywell™ Eclipse 8700 Structural Firefighting Gloves
Gloves have a swing thumb and convertible cuff so it can be worn as a wristlet or a gauntlet style glove
Manufacturer:  Honeywell™ GL8700S
 View more versions of this product
Catalog No. 19-805-40003
Part Not Found
Description
Description
  • Kangaroo outer shell palm stands up to abrasion, and is strong enough for any fire fighting task
  • Densely packed fibers minimize water absorption, and the leather does not become slippery when wet
  • Elk back and anti-slash strip offers superior insulation and high heat stability
  • Pyrotect™ moisture barrier offers very high Moisture Vapor Transmission Rate, excellent low-temperature flexibility, economical breathability
  • Kevlar™/ Nomex™ thermal lining offers heat protection and superior cut resistance
  • Convertible cuff may be worn as wristlet or folded back to convert to gauntlet style
  • 2-D patterning with Swing Thumb™
  • Level of Bloodborne Pathogen Protection: HNO
Compliance
NFPA 1971
Specifications
Specifications
Convertible
Elkskin Leather, Kangaroo Leather
Small
360° Inner pull tab
Dupont™ Kevlar™ aramid fiber, Nomex™ aramid fibers
NFPA 1971