Honeywell Eclipse 8700 Structural Firefighting Gloves
Gloves have a swing thumb and convertible cuff so it can be worn as a wristlet or a gauntlet style glove
Manufacturer: Total Fire Group GL8700XS
 View more versions of this product
Catalog No. 19-805-40002
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Certifications NFPA 1971
    Glove Cuff Style Convertible
    Includes 360° Inner pull tab
    Material Kangaroo, Elkskin Leather
    Liner Material Kevlar™, Nomex™
    Size X-Small
  • Description & Specifications
Related Products
Product Content Feedback