Honeywell Eclipse 8700 Structural Firefighting Gloves
Gloves have a swing thumb and convertible cuff so it can be worn as a wristlet or a gauntlet style glove
Manufacturer: Total Fire Group GL8700LG
 View more versions of this product
Catalog No. 19-805-40005
  Check Availability
Add to Cart
    Specifications
    Certifications NFPA 1971
    Glove Cuff Style Convertible
    Includes 360° Inner pull tab
    Material Kangaroo, Elkskin Leather
    Liner Material Kevlar™, Nomex™
    Size Large
  • Description & Specifications
Related Products
Product Content Feedback