Learn More
Honeywell Fire Warrior Micro Thermal Imager
Small, helmet-mounted, hands-free imager minimizes obstruction of user's vision
Includes: Mounting kit (for use with Morning Pride Ben 2 Plus and Non-Plus Modern Style helmets only), storage case, aluminized training cover, user information guide, and four industrial-grade alkaline “AA” batteries
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
19-130-0369
|
Honeywell
TIMTB1353 |
N/A | Mounting kit for fire warrior micro thermal imager |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Center-mounted thermal imager with adjustable boom lens
- State-of-the-art microbolometer technology for superior visibility with hot spot identification/temperature difference sensing
- High-performance, self-adjusting electronic iris and anti-whiteout programming
- Transfers from fire fighter to fire fighter in seconds
- Low-profile suits even faceshield-equipped helmets; mounting plate is compatible with virtually any brand of helmet1
Order Info
Dimensions: 3 x 3 x 3 in. (7.6 x 7.6 x 7.6cm). Weight: Approximately 1 lb. (0.5kg). Factory-mounting is available for Morning Pride helmets only. Available through Fisher Safety's GSA Advantage! Schedule 84.