Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Honeywell Fire Warrior Micro Thermal Imager
Small, helmet-mounted, hands-free imager minimizes obstruction of user's vision
Manufacturer:  Total Fire Group TIMTB1353
 View more versions of this product
Catalog No. 19-130-0369
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Mounting kit† (for use with Morning Pride Ben 2 Plus and Non-Plus Modern Style helmets only), storage case, aluminized training cover, user information guide, and four industrial-grade alkaline “AA” batteries
Specifications
Description
  • Center-mounted thermal imager with adjustable boom lens
  • State-of-the-art microbolometer technology for superior visibility with hot spot identification/temperature difference sensing
  • High-performance, self-adjusting electronic iris and anti-whiteout programming
  • Transfers from fire fighter to fire fighter in seconds
  • Low-profile suits even faceshield-equipped helmets; mounting plate is compatible with virtually any brand of helmet1
Order Info
Dimensions: 3 x 3 x 3 in. (7.6 x 7.6 x 7.6cm). Weight: Approximately 1 lb. (0.5kg). Factory-mounting is available for Morning Pride helmets only. Available through Fisher Safety's GSA Advantage!™ Schedule 84.