Learn More
Honeywell™ Firemate Gloves
Certified for structural firefighting operations
Manufacturer:  Honeywell™ GL9550L
 View more versions of this product
Catalog No. 19-168-245D
Add to cart
Description
Description
  • Kangaroo outer shell palm stands up to abrasion, slashes, and punctures
  • Densely packed fibers minimize water absorption, prevent leather from becoming slippery when wet
  • Elk back offers superior insulation and high heat stability
  • Crosstech™ moisture barrier provides high breathability, heat resistance, temperature stability, continuous moisture barrier protection, and fast glove drying
  • Thin Kevlar™/ Nomex™ thermal lining offers heat protection and superior cut resistance
  • 2-D patterning with wing thumb with gauntlet wrist
Compliance
Meet NFPA 1971, 2007 Edition.
Specifications
Specifications
Gauntlet
Dupont™ Kevlar™ aramid fiber, Nomex™ aramid fibers
NFPA 1971
Leather, kangaroo
Large