Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Light Task Plus 5™ Cut-Resistant Gloves

Catalog Number p-3742224
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Bi-polymer coated cut-resistant gloves is a blend that combines the best properties of nitrile and PU (polyurethane) to provide a surface that is mechanically tough and flexible.

  • Bi-polymer coating is a blend of nitrile and PU (polyurethane)
  • Comfortable, high performance cut-resistant
  • Seamless form fitting liner contains 'stretch-broken' Dynamo yarn that is cool and flexible
  • 13-gauge liner also contains glass fiber for enhanced cut resistance
  • Knit wrist
TRUSTED_SUSTAINABILITY
Coating Material Bi-Polymer
Color Black
Cut Resistance Rating ANSI A5
Hand Specific
Material Dyneema Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Material handling, assembly work, sheet metal, glass
Glove Cuff Color Orange
Glove Cuff Style Knit
Model NFD20B/7S
Series NFD20B
Yarn Gauge 13
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit