Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Light Task Plus II Cut-Resistant Gloves, Polyurethane Coated

Catalog Number p-3742247
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
Glove Cuff Color:
Blue
Orange
Red
White
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight Dyneema™ cut-resistant fiber gloves with white polyurethane palm and fingertip coating provides cut and abrasion protection.

  • Seamless, cut-resistant liner is made from Dyneema
  • Polyurethane palm coating is light for comfort and maximum tactility
  • High abrasion and puncture resistance
  • Glove conforms to the shape of the hand
  • Back of the glove is uncoated for ventilation and comfort
TRUSTED_SUSTAINABILITY
ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Black, Gray
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Material Dyneema™
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Handling sharp objects, particularly sheet Metal and abrasives handling
Glove Cuff Color White
Glove Cuff Style Knit
Hem Color White
Model NFD15B/10XL
Series NorthFlex Light Task Plus II™
Length (English) 4 in.
Length (Metric) 10.2 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit