Honeywell Flex Light Task Plus II Bi-Polymer Coated Dyneema Gloves

Manufacturer: Honeywell NFD15B/9L

 View more versions of this product

Catalog No. 17-805-002

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    Hand Ambidextrous
    Hem Color Yellow
    For Use With (Application) handling sharp objects, particularly sheet Metal and abrasives handling
    Length (English) 4 in.
    Length (Metric) 10.16cm
    Material Dyneema™
    Size Large
    Coating Material Sandy
    Color Black, gray
    Cut Resistance Rating Level 2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit