Honeywell Flex Nitri Task C5 Foamed Nitrile Palm Coated Aramid/Steel Gloves

Manufacturer: Honeywell NFF13C/8M

 View more versions of this product

Catalog No. 19-130-270B

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    For Use With (Application) sheet Metal fabrication, machine shops, Glass industry, automotive and other assembly work
    Material Aramid, Steel
    Size Medium
    Sterile Yes
    Coating Material Seamless
    Color Dark blue
    Cut Resistance Rating Level 4

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit