Honeywell Grip-Task Series Natural Rubber Gloves

Protect against abrasion, liquids and chemicals

Manufacturer: Honeywell T66FWG/11XXL

 View more versions of this product

Catalog No. 19-150-0961

  Check Availability
Add to Cart

Alert

This product contains natural rubber latex proteins which may cause allergic reactions in some individuals.


    Specifications

    Exterior Finish Wrinkle
    Glove Cuff Style Safety
    For Use With (Application) chemical, construction, manufacturing, municipal services
    Length (English) 12 in.
    Length (Metric) 30cm
    Material Natural Rubber
    Liner Material Cotton Jersey
    Size 2X-Large
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit