Honeywell Junkyard Dog Seamless Knit with Leather Palm Gloves
Provide excellent cut and abrasion protection
Manufacturer: Honeywell KV18AL10050
 View more versions of this product
Catalog No. 19-049-941
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) automotive assembly, sheet Metal, Glass handling, Metal stamping, Metal fabrication, recycling, canning, injection moulding, Paper/pulp industry
    Material Kevlar™
    Size Women's
    Weight Medium weight
    Color Yellow
    Cut Resistance Rating Level 3
  • Description & Specifications
Related Products
Product Content Feedback