Learn More
Honeywell™ Kangaroo Leather Super Gloves
Provide superior comfort and durability
Manufacturer:  Honeywell™ GLSGKCGXL
 View more versions of this product
Catalog No. 19-171-019E
Add to cart
Description
Description
  • Top-grain kangaroo leather on glove back offers easy flex and exceptional durability
  • Black Digiroo digital kangaroo grip palms and fingers for incredible durability, grip and tactility
  • Bubble-Flex and Flex-Tuck construction with dead-air spacer ridges allow natural hand flexing
  • True 3-D styling with staggered layer seaming ensures wearer comfort
  • Air Spacer thermal architecture traps air for thermal protection without traditional bulk
  • Durable, high-temperature-stable CrossTech™ moisture barrier and front and back black eversoft cowhide cuffs
Compliance
Meet NFPA™ 1971, 2007 Edition.
Specifications
Specifications
Gauntlet
Dupont™ Kevlar™ aramid fiber, Nomex™ aramid fibers, Thermal
NFPA 1971
Leather, kangaroo
X-Large