Learn More
Honeywell™ Aramid Fiber Sleeve
Manufacturer:  Honeywell™ KVCS218TH
Catalog No. 19-816-579
Add to cart
Specifications
Specifications
Level 3
Metal sheet handling, recycling, automotive assembly, blade handling, knife sharpening, Glass handling
Medium weight
18 in.
Knit
Dupont™ Kevlar™ aramid fibers
Yellow
45.72cm