Honeywell Lockout/Tagout Kits

Manufacturer: Honeywell LK108FE

 View more versions of this product

Catalog No. 19-170-151

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Manufacturing, mining, municipal services, oil and gas, utilities
    Color Yellow
    Includes Nylon carrying bag, circuit breaker lockouts, electrical switch lockouts, electrical plug lockouts, nylon lockout hasps, metal lockout hasps, padlocks, 10 lockout tags
    Product Type Electrical Lockout/Tagout Kit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit