Learn More
Honeywell Long Sleeve PVC Gloves
Manufacturer:  Honeywell 1812FWG01
 View more versions of this product
Catalog No. 19-150-1013
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Rough
Ambidextrous
Interlock knit
Large
Green
66.04cm
Suspender Strap
agriculture, chemical and food services
PVC (Polyvinyl Chloride)
ANSI/ISEA 105, ASTM D 3389-05, ANSI/ISEA 105, ASTM F 1790-97, EN 374, EN 388, EN 420, OSHA 29 CFR 1915.157
26 in.