Honeywell LTO-3403; Royal Blue Proximity Gear, Beaufort MCAS Fire Dept. SC Specs Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Total Fire Group GSAMCA00137

 View more versions of this product

Catalog No. 19-300-306

  Check Availability
Add to Cart

    Specifications

    Color Royal Blue
    For Use With (Equipment) LTO 2403
    Product Type Tails Coat

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit