Honeywell MaskMate Hoods Available on GSA/VA Contract for Federal Government customers only.

Universal fit to all brands of air masks

Manufacturer: Total Fire Group HDCKCKSQ

 View more versions of this product

Catalog No. 19-823-193

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance NFPA 1971
    Quantity Each
    Includes Square bib, notched shoulders, P84 blend
    Size Universal
    For Use With (Equipment) Universal fit to all brands of air mask
    Resistance Flame
    Color Black Carbon

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit