Honeywell Milan Laser Eyewear
Polymer filters feature fully integrated dyes for superior protection
Manufacturer: Honeywell 3160103
 View more versions of this product
Catalog No. 18-813-307
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance ANSI Z87.1-2003 and ANSI Z136.1 standards
    Filter Type 103
    Lens Tint Orange
    Surface Coating Antifog, antiscratch
    Type Laser Eyewear
    Color Orange
    Transmittance Value 50%
  • Description & Specifications
Related Products
Product Content Feedback