Learn More
Honeywell™ Mururoa V4F1 Ventilated Protective Suits
Ventilated protective suit against radioactive contamination.
Manufacturer:  Honeywell™ CC8481413/DEL
 View more versions of this product
Catalog No. 18-001-052
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Made of pink Flame-Retardant M1, PVC 20/100mm 
  • High protection factor (class 5 according to EN 1073-1)
  • Has to be plugged to breathable air network 5,5 Bar dynamic
Specifications
Specifications
EN943-1, type 1c, EN 1073-1, NFP 92507
T3
Simple and convenient air regulation: air flow adjustable with a valve, 360° field of vision, Low noise level, Built-in ventilation
Yes
Fabric
Pink
Nuclear, chemical and pharmaceutical industries in areas
Class 5 (>50,000)
PVC 20/100th flame retardant M1; T3, gloves size 9