Learn More
Honeywell NitriGuard Nitrile Gloves
With sanitized interior
Manufacturer:  Honeywell LA225G/8
 View more versions of this product
Catalog No. 19-150-1047
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Sand patch
chemical and food industry, laboratories, cleaning and other applications requiring flexibility and dexterity
Nitrile
22mil
Green
38.1cm
Straight
Flock
8
21 CFR, USDA, FDA
15 in.