Honeywell NitriTask Supported Foam Nitrile Gloves

Versatile cut-resistant work gloves

Manufacturer: Honeywell NFF13/9L

 View more versions of this product

Catalog No. 19-130-1658

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Launderable
    Glove Cuff Style Knit
    For Use With (Application) automotive assembly, general maintenance work
    Length (English) 1 in.
    Length (Metric) 2.54cm
    Liner Material White Nylon
    Color Gray
    Cut Resistance Rating Level 1
    Material Nitrile
    Size 9

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit