Learn More
Honeywell™ Nomex Flight Gloves
Offer excellent protection from flash and flame hazard and exceed USAF specifications for thermal protection
Manufacturer:  Honeywell™ 161010
 View more versions of this product
Catalog No. 19-816-629
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Manufactured to U.S. military specifications, these gloves are exceptionally supple for optimal dexterity.
  • Made from 100% Nomex™ brand fiber Grade A, chrome-tanned, sheepskin leather palms Color: Sage green
Order Info
Selected sizes are featured in ordering table.
Compliance
Exceed USAF specifications for thermal protection.
Specifications
Specifications
flash and flame hazard
10
USAF
Leather
Sage Green