Learn More
Honeywell™ Nomex™ Flight Gloves
Gloves offer protection against flash and flame hazards, in addition to exceeding USAF specifications for thermal protection. The longer cuff length is designed to extend past jacket cuffs to ensure that wrists remain protected at all times.
Manufacturer: Honeywell™ 161011
Description
- Constructed from 100% DuPont™ Nomex™ Grade A fibers for flame and heat resistance
- Chrome-tanned, sheepskin leather palms enhance grip capabilities
- Gloves exceed all USAF specifications for thermal protection
- Manufactured to U.S. military specifications
- Exceptionally supple for optimal dexterity
Compliance
Exceeds USAF Specifications for Thermal Protection
Specifications
Green, Gray | |
Unisex | |
Nomex™ | |
1610 | |
Exceeds USAF Specifications for Thermal Protection | |
Manufactured to meet US Military Specifications |
Flight Operations, and Piloting | |
Open | |
Sheepskin Leather | |
11 | |
Specific |