Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Nomex Flight Gloves
Offer excellent protection from flash and flame hazard and exceed USAF specifications for thermal protection
Specifications
Certifications/Compliance | USAF |
---|---|
Color | Sage Green |
Material | Leather |
For Use With (Application) | flash and flame hazard |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-816-629
|
Honeywell
161010 |
10 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-816-628
|
Honeywell
16109 |
9 |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-816-630
|
Honeywell
161011 |
11 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Manufactured to U.S. military specifications, these gloves are exceptionally supple for optimal dexterity.
- Made from 100% Nomex brand fiber Grade A, chrome-tanned, sheepskin leather palms Color: Sage green
Order Info
Selected sizes are featured in ordering table.
Compliance
Exceed USAF specifications for thermal protection.
Specifications
USAF | |
Sage Green | |
Leather | |
flash and flame hazard |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title