Honeywell NorthFlex™ Neon Hi-Viz Foamed PVC Coated Nylon Gloves

Multi-purpose gloves in Hi-Viz yellow color for low-light safety

Manufacturer: Honeywell NF11HVY/8M

 View more versions of this product

Catalog No. 17-805-006

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Launderable
    Glove Cuff Style Knit
    For Use With (Application) construction, municipal services, transportation
    Material PVC
    Liner Material Nylon
    Size Medium
    Coated Area Palm
    Color Hi-vis yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit