Learn More
Honeywell NorthFlex™ Nitri Task KL Gloves
Comfortable, dexterous cut-resistant work gloves
Manufacturer:  Honeywell NFKL13/6XS
 View more versions of this product
Catalog No. 19-150-0992
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Palm, finger
Textured
cut and abrasion resistance without compromising dexterity, tactility and comfort
Dyneema™
Green
Level 2
Knit
Nitrile
X-Small
Black,Yellow