Honeywell PBI Aluminized LTO Proximity Pants
For applications where sustained operations in high heat are anticipated
Manufacturer: Total Fire Group SAEGSA00478MD
 View all options for this product
Catalog No. 19-165-759
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Color Silver
    Size Medium
    Material Aluminized PBI Silver; Two-layer Spunlace/meta-aramid Thermal Liner; CrossTech™/Nomex™ Moisture Barrier
    • Outer shell of 7 oz. aluminized PBI silver
    • Two-layer spunlace/meta-aramid thermal liner
    • Detachable inner liner
    • CrossTech™/Nomex™ moisture barrier
    • 1.5 in. Velcro™ fly
    • Angled cuffs
    • Cushioned knees
    • Two take-up postman straps
    • Bellows pockets: 9 x 9 x 1 1/2 in.
    • Navy NFPA 2007 Edition Specifications
  • Description & Specifications
Related Products
Product Content Feedback